Orders LinkContactUsLinkComponent
Assortment Boxes, PP
Assortment Boxes, PP
Assortment box, with hinged lid and 4×8 compartments, Anthracite, 340×260×55 mm
Catalog # 215-0289
Supplier:   SEMADENI
Assortment Boxes, PP
Catalog # 215-0289
Supplier:  SEMADENI
Assortment Boxes, PP
Assortment Boxes, PP
Assortment box, with hinged lid and 4×8 compartments, Anthracite, 340×260×55 mm
Price
60,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
  • Robust and strong design
  • All models can be stacked together
  • Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
Specifications
  • Colour:
    Anthracite
  • Description:
    Assortment box, with hinged lid and 4×8 compartments
  • Seq:
    06
  • W×D×H:
    340×260×55 mm