ContactUsLinkComponent
TICKOPUR R 36 - 5 LITRES, SPECIAL CLEANER, FOR THE ANALYTICAL APPLICATION, LASER TECHNOLOGY AND BLADE CLEANING, TENSIDE-FREE, CONCEN TRATE, NON-FOAMING, GENTLE TO MATERIAL, DOSAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 5 L
  142-1921
TICKOPUR R 36 - 5 LITRES, SPECIAL CLEANER, FOR THE ANALYTICAL APPLICATION, LASER TECHNOLOGY AND BLADE CLEANING, TENSIDE-FREE, CONCEN TRATE, NON-FOAMING, GENTLE TO MATERIAL, DOSAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 5 L
  142-1921
Frequently Bought Together