Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Glass bottle for solids, 500 g
Catalog # 40476.36
Oxidizing
Irritant
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Catalog # 40476.36
CAS Number:  1304-85-4
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Glass bottle for solids, 500 g
CAS Number:  1304-85-4
Price
277,00 €
Log in for availability
1 * 500 g
Product Details & Documents
CAS No.: 1304-85-4 Molecular Formula: Bi5O(OH)9(NO3)4 (approximate) Formula Weight: 1461.98 Physical Form: Powder Density: 6.04 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 260° dec. Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl Merck Reference: 14,1284 UN No.: 1477 MDL No.: MFCD00064844
Ceramic glazes and enamel fluxes
Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Merck Index:14,1284
  • Melting Pt:260 °C (dec.)
  • Density:6.04
  • Storage Temperature:Ambient
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g