Boron phosphate
500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 13308-51-5 Molecular Formula: BPO4 Formula Weight: 105.79 Physical Form: Powder Density: 2.81 Appearance: White Odour: Odourless Melting Point: 1400° MDL No.: MFCD00011318
- CAS Number:13308-51-5
- Formula:BO₄P
- MDL Number:MFCD00011318
- Molecular Weight:105.78 g/mol
- Melting Pt:>1400 °C
- Density:2.81
- Storage Temperature:Ambient
- EINECS:236-337-7
Documents
Specifications
Specifications
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



