Orders LinkContactUsLinkComponent
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Spare 80 ml grinding chamber, Low temperature applications (-200 °C). ETFE and reinforced glass-fibre with stainless steel inlet, has good chemical resistance
Catalog # 412-0001
Supplier:   IKA
Accessories for analytical Mill, A 11 basic
Catalog # 412-0001
Supplier:  IKA
Accessories for analytical Mill, A 11 basic
Accessories for analytical Mill, A 11 basic
Spare 80 ml grinding chamber, Low temperature applications (-200 °C). ETFE and reinforced glass-fibre with stainless steel inlet, has good chemical resistance
Price
545,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Spare 80 ml grinding chamber
  • Seq:
    01
  • For:
    Low temperature applications (-200 °C). ETFE and reinforced glass-fibre with stainless steel inlet, has good chemical resistance