Inoculating loops and needles, disposable
Loop, 1 µl, White, 50 loops per sleeve, 40 sleeves per case
Catalog # 612-9300
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
Flexible polymer.
- Sterilised by irradiation
- Individually wrapped
- Length: 200 mm
Certificates
Search by Lot NumberSpecifications
- Packed:50 loops per sleeve, 40 sleeves per case
- Capacity:1 µl
- Colour:White
- Description:Loop
- Seq:02