Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Loop, 1 µl, White, 50 loops per sleeve, 40 sleeves per case
Catalog # 612-9300
Supplier:   Greiner Bio-One
Inoculating loops and needles, disposable
Catalog # 612-9300
Supplier:  Greiner Bio-One
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Loop, 1 µl, White, 50 loops per sleeve, 40 sleeves per case
Price
212,00 €
Log in for availability
1 * 2.000 items
Product Details & Documents
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
Specifications
  • Packed:
    50 loops per sleeve, 40 sleeves per case
  • Capacity:
    1 µl
  • Colour:
    White
  • Description:
    Loop
  • Seq:
    02