Orders LinkContactUsLinkComponent
Phase separating paper, grade 1PS
90 mm
  515-0086
Phase separating paper, grade 1PS
  515-0086
Phase separating paper, grade 1PS
Phase separating paper, grade 1PS
90 mm
56,50 €
1 * 100 items
1PS phase-separating paper is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
  • Impregnated with stabilised silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the 1PS phase-separating paper. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
  • Seq:
    02
  • Size:
    90 mm