Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, pointed, 145 mm
Catalog # 232-1913
Supplier:   VITLAB
Forceps
Catalog # 232-1913
Supplier:  VITLAB
Forceps
Forceps
Straight, pointed, 145 mm
Price
2,20 €
Log in for availability
1 * 1 items
Product Details & Documents
PMP, white. Multipurpose forceps for a wide range of applications.
  • Good chemical resistance
  • Autoclavable
  • Robust, pliable, self-sprung
Specifications
  • Type:
    Straight, pointed
  • Length:
    145 mm