Spray bottles
850 ml, Spray bottle, White
VITL53610
: VITLAB
White or translucent made from PP.
- Excellent chemical resistance
- Excellent temperature resistance
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet. Range: approx. 3 to 4 metres. Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.
- Capacity:850 ml
- Colour:White
- Description:Spray bottle
- Seq:02