Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
185 mm, 5 mm
Catalog # 231-2152
Supplier:   RSG
Spoon spatulas, micro
Catalog # 231-2152
Supplier:  RSG
Spoon spatulas, micro
Spoon spatulas, micro
185 mm, 5 mm
Price
4,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • With round handle and one bent spoon
  • Excellent temperature resistance, autoclavable
  • Good resistance to chemicals
  • Very flexible
Documents
Specifications
  • Length:
    185 mm
  • Blade width:
    5 mm