Orders LinkContactUsLinkComponent
Double spatulas, narrow
Double spatulas, narrow
Stiff, 210 mm, 11 mm
Catalog # RSGA830.210
Supplier:   RSG
Double spatulas, narrow
Catalog # RSGA830.210
Supplier:  RSG
Double spatulas, narrow
Double spatulas, narrow
Stiff, 210 mm, 11 mm
Price
36,80 €
Log in for availability
1 * 5 items
Product Details & Documents
Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.
Specifications
  • Description:
    Stiff
  • Length:
    210 mm
  • Blade width:
    11 mm