Orders LinkContactUsLinkComponent
Tripods
Tripods
With detachable legs, 180×210 mm
Catalog # RSGA493.180
Supplier:   RSG
Tripods
Catalog # RSGA493.180
Supplier:  RSG
Tripods
Tripods
With detachable legs, 180×210 mm
Price
69,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Separable tripods made from stainless steel.
Specifications
  • Int.Ø×H:
    180×210 mm
  • Description:
    With detachable legs
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Tripod extensions, set of three, total height up to 370 mm
    Tripod extensions, set of three, total height up to 370 mm
    RSGA497.370
    Log in for availability
    1 * 3 items - 78,80 €