Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 145 mm
Catalog # 232-2166
Supplier:   RSG
Forceps with guide pin
Catalog # 232-2166
Supplier:  RSG
Forceps with guide pin
Forceps with guide pin
Curved, with guide pin, 145 mm
Price
13,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel, anti-magnetic. Particularly suitable for staining applications.
  • With curved, extra fine tips
  • Corrosion resistant
Specifications
  • Type:
    Curved, with guide pin
  • Length:
    145 mm