Safety canisters
Safety canister with self-closing tap 1½" and sep. ventilation, 20 l
Catalog # 119-2380
Supplier: ROETZMEIER SICHERH BEHAELTER
Jump to:
Product Details & Documents
Stainless steel.
- In-plant use of flammable liquids, such as solvents
- Explosion-proof due to flame traps (removable) and pressure control valves
- Taps and metering devices are self-closing
- High quality, chemically resistant stainless steel (0,8 mm thick) with PTFE gaskets
- Easy handling due to ergonomic design
- Models with self-closing taps are equipped with a separate ventilation screw and are suitable for horizontal storage and decanting
Canister shape maximises the usage of limited storage space, e.g. in safety cabinets.
Specifications
- Capacity:20 l
- Description:Safety canister with self-closing tap 1½" and sep. ventilation
- Seq:09
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Flame trapFlame trap119-0169All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)Log in for availability1 * 1 items - 25,10 €
Product Information
- Catalog #Ø×HForAvailabilityPrice
- Rotable spout 1½“Rotable spout 1½“119-016128×175 mmAll Rötzmeier safety canisters and barrels (10 and 25 l)Log in for availability1 * 1 items - 81,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Screw cap 1½" with pressure control valveScrew cap 1½" with pressure control valve119-0129119-0172, -0116, -2380, -2045, -2046, -0121, -2180, -2181, -2182, -2160, -0111, -0115, -0123Log in for availability1 * 1 items - 63,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Self-closing tap 1½"Self-closing tap 1½"119-2401119-2180, -2181, -2182, -0122, -0157, -0156Log in for availability1 * 1 items - 112,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Self-closing metering device 1½“ for safety canisters and barrelsSelf-closing metering device 1½“ for safety canisters and barrels119-2391119-0172, -0116, -2045, -2046, -2370, -2371, -0115Log in for availability1 * 1 items - 87,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



