Orders LinkContactUsLinkComponent
Hoffmann pinch cocks
Hoffmann pinch cocks
50 mm
Catalog # 229-0347
Supplier:   USBECK Laborgeräte
Hoffmann pinch cocks
Catalog # 229-0347
Hoffmann pinch cocks
Hoffmann pinch cocks
50 mm
Price
22,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Nickel plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Hinged
  • Very good thermal resistance
  • The standard model has a knurled screw and the heavy duty model has a butterfly screw
  • Easy cleaning
Ordering information: The values in the band width column refer to the clearance between the pinch cock legs.
Specifications
  • Bandwidth:
    50 mm