Orders LinkContactUsLinkComponent
Scoops
Scoops
1500 ml, 390 mm
Catalog # USBE3726
Supplier:   USBECK Laborgeräte
Scoops
Catalog # USBE3726
Scoops
Scoops
1500 ml, 390 mm
Price
29,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    1500 ml
  • Length:
    390 mm