Orders LinkContactUsLinkComponent
Scoops
Scoops
130 ml, 210 mm
Catalog # 231-0324
Supplier:   USBECK Laborgeräte
Scoops
Catalog # 231-0324
Scoops
Scoops
130 ml, 210 mm
Price
5,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    130 ml
  • Length:
    210 mm