Orders LinkContactUsLinkComponent
Stand rods
12 mm, 750 mm
Catalog # 241-7162
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # 241-7162
Stand rods
Stand rods
12 mm, 750 mm
Price
9,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Steel, galvanised.
  • Good chemical resistance
  • Very sturdy
Specifications
  • Ø:
    12 mm
  • Length:
    750 mm