Orders LinkContactUsLinkComponent
Stand rods
Stand rods
12 mm, Steel rod, 880 g, 1000 mm, M 10
Catalog # USBE2004/2104
Supplier:   USBECK Laborgeräte
Stand rods
Catalog # USBE2004/2104
Stand rods
12 mm, Steel rod, 880 g, 1000 mm, M 10
Price
46,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Steel, zinc plated.
  • Threaded (M 10)
  • Diameter: 12 mm
Specifications
  • Ø:
    12 mm
  • Thread:
    M 10
  • Description:
    Steel rod, 880 g
  • Length:
    1000 mm
  • Seq:
    04