Orders LinkContactUsLinkComponent
Plates and modules, FluoroNunc™ and LumiNunc™
Plates and modules, FluoroNunc™ and LumiNunc™
MaxiSorp™, F16×6 wells, black*, Black, 350 µl
Catalog # 735-0070
   Low Manufacturing Impact
Plates and modules, FluoroNunc™ and LumiNunc™
Catalog # 735-0070
   Low Manufacturing Impact
Plates and modules, FluoroNunc™ and LumiNunc™
Plates and modules, FluoroNunc™ and LumiNunc™
MaxiSorp™, F16×6 wells, black*, Black, 350 µl
Price
356,00 €
Log in for availability
1 * 60 items
Product Details & Documents
PS, with MaxiSorp™ or PolySorp™ treated surface. For fluorescence and luminescence based immunoassays and binding assays. Available with a choice of certified MaxiSorp™ surface, with high affinity to molecules with mixed hydrophilic/hydrophobic domains, or, PolySorp™ surface, less polar than MaxiSorp™ and with affinity to molecules of a more hydrophobic nature.
  • Choice of framed modules or 96-well plates with either flat (F) or C-shaped wells
  • White modules/plates give maximum reflection with minimum autofluorescence and autoluminescence
  • Black modules/plates give minimum background in fluorescence reading, allow minimum back scattered light, and minimise cross talk
  • Clear modules/plates are suitable for time-resolved fluorescence
* With certificate
Specifications
  • Recommended working volume:
    350 µl
  • Colour:
    Black
  • Description:
    MaxiSorp™, F16×6 wells, black*
  • Seq:
    04
  • Cat. No.:
    735-0070
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact