Plates and modules, FluoroNunc™ and LumiNunc™
PolySorp™, F96, white, White, 350 µl, 96
Catalog # 735-0003
Supplier: Thermo Fisher Scientific
Plates and modules, FluoroNunc™ and LumiNunc™
Catalog # 735-0003
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PS, with MaxiSorp™ or PolySorp™ treated surface. For fluorescence and luminescence based immunoassays and binding assays. Available with a choice of certified MaxiSorp™ surface, with high affinity to molecules with mixed hydrophilic/hydrophobic domains, or, PolySorp™ surface, less polar than MaxiSorp™ and with affinity to molecules of a more hydrophobic nature.
- Choice of framed modules or 96-well plates with either flat (F) or C-shaped wells
- White modules/plates give maximum reflection with minimum autofluorescence and autoluminescence
- Black modules/plates give minimum background in fluorescence reading, allow minimum back scattered light, and minimise cross talk
- Clear modules/plates are suitable for time-resolved fluorescence
* With certificate
Specifications
- No. of wells:96
- Recommended working volume:350 µl
- Colour:White
- Description:PolySorp™, F96, white
- Seq:11
- Cat. No.:735-0003
- Environmental certification:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact