Extraction thimbles
48×200 mm, 2,0 mm
Catalog # 515-8949
Supplier: Whatman products (Cytiva)
Jump to:
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Specifications
- Int.Ø×H:48×200 mm
- Seq:29
- Thickness:2,0 mm