Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
27×80 mm, 1,5 mm
Catalog # 533-0004
Extraction thimbles
Catalog # 533-0004
Extraction thimbles
Extraction thimbles
27×80 mm, 1,5 mm
Price
106,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Specifications
  • Int.Ø×H:
    27×80 mm
  • Seq:
    08
  • Thickness:
    1,5 mm