Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Cap Inserts
CapInsert™ cap inserts for Cryovial® tubes
CapInsert™ cap inserts for Cryovial® tubes
Print
CapInsert™ cap inserts for Cryovial® tubes
White
Catalog # 479-4531
Supplier:
Simport Scientific
CapInsert™ cap inserts for Cryovial® tubes
Catalog # 479-4531
Supplier:
Simport Scientific
Expand image
8
Return to Top
CapInsert™ cap inserts for Cryovial® tubes
White
Specifications
Add to cart
Jump to:
Specifications
Price
37,50 €
Log in
for availability
Unit of Measure
1 * 500 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
CoC_SIMPT312-10_C19282632
Specifications
Colour:
White
Seq:
11
VWR® NITRILE LIGHT, Nitrile Gloves
#: 112-2756
Supplier: VWR Collection