Embedding cassettes, M480
White
720-0832
Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures.
- Resistant to all commonly used histological solvents
- Designed to accept standard metal lids (M481)
- Slanted writing surface (45° angle) accepts markings easily
Different colours available.
- Colour:White
- Seq:05