Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
150 mm, 4×6×1,4 mm, 0,1 ml
Catalog # 231-0171
Supplier:   NICKEL ELECTRO
Spoon spatulas, micro
Catalog # 231-0171
Supplier:  NICKEL ELECTRO
Spoon spatulas, micro
Spoon spatulas, micro
150 mm, 4×6×1,4 mm, 0,1 ml
Price
139,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Stainless steel.
  • One end flattened, the other end spoon-formed
  • Good chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Specifications
  • Capacity:
    0,1 ml
  • Length:
    150 mm
  • Spoon size:
    4×6×1,4 mm