Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
Plug cable combination, LB 3 A (coaxial electrode plug (for BlueLine), 3 m coaxial cable, DIN instrument plug), 3 m
Catalog # 662-2193
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2193
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
Plug cable combination, LB 3 A (coaxial electrode plug (for BlueLine), 3 m coaxial cable, DIN instrument plug), 3 m
Price
80,00 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Plug cable combination, LB 3 A (coaxial electrode plug (for BlueLine), 3 m coaxial cable, DIN instrument plug)
  • Length:
    3 m
  • Seq:
    23