Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
Plug cable combination, LB 1 BNC (Coaxial electrode plug (for BlueLine), 1 m coaxial cable, BNC instrument plug), 1 m
Catalog # 662-6165
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-6165
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
Plug cable combination, LB 1 BNC (Coaxial electrode plug (for BlueLine), 1 m coaxial cable, BNC instrument plug), 1 m
Price
66,00 €
Log in for availability
1 * 1 items
Specifications
  • Description:
    Plug cable combination, LB 1 BNC (Coaxial electrode plug (for BlueLine), 1 m coaxial cable, BNC instrument plug)
  • Length:
    1 m
  • Seq:
    22