Orders LinkContactUsLinkComponent
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
2000 ml, 29/32 NS
Catalog # SCOT242946308
Supplier:   DWK Life Sciences
Conical separating funnels, DURAN®
Catalog # SCOT242946308
Supplier:  DWK Life Sciences
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
2000 ml, 29/32 NS
Price
201,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml