Separating funnels, spherical, DURAN®
500 ml
Catalog # 527-1412
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3, clear, stopcock with glass key and plastic stopper.
- Without graduation
- Very good chemical resistance
- Very good temperature resistance
Specifications
- Capacity:500 ml
- Cat. No.:527-1412