Orders LinkContactUsLinkComponent
Specimen loops
Specimen loops
Specimen loop, platinum-iridium, 0,7 mm, 1 mm, 40 mm
Catalog # 631-0839
Supplier:   KLS Martin Group
Specimen loops
Catalog # 631-0839
Supplier:  KLS Martin Group
Specimen loops
Specimen loop, platinum-iridium, 0,7 mm, 1 mm, 40 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Ø:
    1 mm
  • Description:
    Specimen loop, platinum-iridium, 0,7 mm
  • Length:
    40 mm