Tanks, rectangular, with cover, Nalgene®
8 L, 200×200×200 mm
Catalog # 216-1921
Supplier: Thermo Fisher Scientific
Tanks, rectangular, with cover, Nalgene®
Catalog # 216-1921
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
- Good stress-crack resistance
- Suitable for use with many organic chemicals
- Tanks can be modified with a factory-installed spigot
Specifications
- Capacity:8 L
- Seq:01
- W×D×H:200×200×200 mm
- Cat. No.:216-1921
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact