Test tubes, flat bottom
65 mm, 15 mm, 0,8 mm
Catalog # SCERB406515000812
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Specifications
- Ext.Ø:15 mm
- Wall thickness:0,8 mm
- Length:65 mm
- Seq:05