Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
9-10 mm
Catalog # LUDI184111411
Supplier:   LUDI
Caps, LABOCAP, without grip
Catalog # LUDI184111411
Supplier:  LUDI
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
9-10 mm
Price
21,20 €
Log in for availability
1 * 10 items
Product Details & Documents
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Specifications
  • For Ø ext.:
    9-10 mm
  • Seq:
    01