Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
17 - 18 mm, Blue
  391-5911
 :   LUDI
Caps, LABOCAP, without grip
  391-5911
 :  LUDI
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
17 - 18 mm, Blue
20,10 €
1 * 10 items
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
  • For Ø ext.:
    17 - 18 mm
  • Colour:
    Blue
  • Seq:
    13