Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
15-16 mm, Blue
Catalog # 391-5908
Supplier:   LUDI
Caps, LABOCAP, without grip
Catalog # 391-5908
Supplier:  LUDI
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
15-16 mm, Blue
Price
19,40 €
Log in for availability
1 * 10 items
Product Details & Documents
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
Specifications
  • For Ø ext.:
    15-16 mm
  • Colour:
    Blue
  • Seq:
    09