Caps, LABOCAP, without grip
15-16 mm, Red
391-5909
: LUDI
Made of anodised aluminum with stainless steel spring clip.
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
- For Ø ext.:15-16 mm
- Colour:Red
- Seq:08