Caps, LABOCAP, without grip
15 - 16 mm, Silver
391-5907
: LUDI
Made of anodised aluminum with stainless steel spring clip.
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
- For Ø ext.:15 - 16 mm
- Colour:Silver
- Seq:07