Orders LinkContactUsLinkComponent
Caps, LABOCAP, without grip
12 - 13 mm, Silver
  391-5901
 :   LUDI
Caps, LABOCAP, without grip
  391-5901
 :  LUDI
Caps, LABOCAP, without grip
Caps, LABOCAP, without grip
12 - 13 mm, Silver
18,10 €
1 * 10 items
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
  • For Ø ext.:
    12 - 13 mm
  • Colour:
    Silver
  • Seq:
    02