Orders LinkContactUsLinkComponent
Swabs
Swabs
Non sterile swabs, Wood/cotton, 2,2 mm, 150 mm
Catalog # LPIT112191
Supplier:   LP ITALIANA
Swabs
Catalog # LPIT112191
Supplier:  LP ITALIANA
Swabs
Swabs
Non sterile swabs, Wood/cotton, 2,2 mm, 150 mm
Price
113,00 €
Log in for availability
1 * 3.000 items
Product Details & Documents
Sterile and non sterile swabs designed for use in microbiology.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Ø:
    2,2 mm
  • Description:
    Non sterile swabs
  • Length:
    150 mm
  • Shaft/head:
    Wood/cotton