Orders LinkContactUsLinkComponent
Adapters, delivery, angled
Adapters, delivery, angled
19/26 NS, 200 mm
Catalog # 201-0366
Supplier:   Lenz Laborglas
Adapters, delivery, angled
Catalog # 201-0366
Supplier:  Lenz Laborglas
Adapters, delivery, angled
Adapters, delivery, angled
19/26 NS, 200 mm
Price
20,80 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint socket and delivery tube at an angle of 105°. Available with long and short delivery tubes.
Documents
Specifications
  • Socket size:
    19/26 NS
  • Tube length:
    200 mm