Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
Catalog # LENZ05000414
Supplier:   Lenz Laborglas
Multiple adapters
Catalog # LENZ05000414
Supplier:  Lenz Laborglas
Multiple adapters
Multiple adapters
29/32 NS, 29/32 NS
Price
33,10 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With two standard ground 29/32 sockets and one standard ground 29/32 cone. Side neck inclined upwards at an angle of 105°.
Documents
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS
  • Seq:
    03