Orders LinkContactUsLinkComponent
Bath fluids
Bath fluids
Kryo 30, –30...+90 °C, − °C, − °C, Light yellow, 10 l
Catalog # 461-2864
Supplier:   LAUDA
Bath fluids
Catalog # 461-2864
Supplier:  LAUDA
CAS Number:  
Bath fluids
Bath fluids
Kryo 30, –30...+90 °C, − °C, − °C, Light yellow, 10 l
CAS Number:  
Price
318,00 €
Log in for availability
1 * 10 L
Product Details & Documents
Allow highly accurate thermostating, even at extreme temperatures.
  • Simple and safe handling, suitable for long term operation
  • Optimise thermostat life
* Denotes Silicone oil
** Only suitable for closed circuits
Specifications
Specifications
  • Pk:
    10 l
  • Colour:
    Light yellow
  • Fire point:
    − °C
  • Flash point:
    − °C
  • Temperature range:
    –30...+90 °C
  • Type:
    Kryo 30
Specifications