Large Dewar flasks, type C/CAL
Glass refill, type 40, Large Dewar flask, 30/4 C and 30/4 CAL
Catalog # 478-0734
Supplier: KGW
Jump to:
Product Details & Documents
Dewar flasks made of borosilicate glass 3.3 DIN/ISO 3585, for storing and tranporting of LN₂, CO₂ and other cooling liquids. Suitable as insulating flasks.
- With handle or two grips
- Insulated lid (incl. grip)
- The Dewar flasks are vacuum insulated and silvered
- Large aperture
- Robust design, with quick release clamp
Type C= metal - blue coated
Type CAL= Stucco aluminium
Temperature range: −200 to +200 °C.
Specifications
- Description:Glass refill, type 40
- Seq:15
- For:Large Dewar flask, 30/4 C and 30/4 CAL