Orders LinkContactUsLinkComponent
Screw caps for Jerricans
60 mm, 20, 25 and 30 L jerricans
Catalog # 216-4312
Supplier:   Kautex
Screw caps for Jerricans
Catalog # 216-4312
Supplier:  Kautex
Screw caps for Jerricans
Screw caps for Jerricans
60 mm, 20, 25 and 30 L jerricans
Price
1,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • For:
    20, 25 and 30 L jerricans
  • Thread:
    60 mm
  • Seq:
    21