Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Screw caps for Jerricans

Screw caps for Jerricans

60 mm, 30 L jerricans
Catalog # 216-5569
Supplier:   Kautex
Screw caps for Jerricans
Catalog # 216-5569
Supplier:  Kautex
Screw caps for Jerricans
Screw caps for Jerricans

60 mm, 30 L jerricans

Price
80,40 €
Log in for availability
1 * 1 items
Product Details & Documents

Documents

Specifications
  • For:
    30 L jerricans
  • Thread:
    60 mm
  • Seq:
    29
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.