Orders LinkContactUsLinkComponent
Racks, Plexiglas for tubes
Racks, Plexiglas for tubes
12 (2×6), 40 mm, 310×110×80 mm
Catalog # 212-0223
Supplier:   KARTELL
Racks, Plexiglas for tubes
Catalog # 212-0223
Supplier:  KARTELL
Racks, Plexiglas for tubes
Racks, Plexiglas for tubes
12 (2×6), 40 mm, 310×110×80 mm
Price
72,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Plexiglas (PMMA)
  • Two tiers
  • Smaller holes in the lower tier to hold test tubes securely
Specifications
  • For tube Ø:
    40 mm
  • No. of Holes:
    12 (2×6)
  • W×D×H:
    310×110×80 mm