Orders LinkContactUsLinkComponent
Filter discs
Filter discs
240 mm
Catalog # 519-5008
Supplier:   KARTELL
Filter discs
Catalog # 519-5008
Supplier:  KARTELL
Filter discs
Filter discs
240 mm
Price
18,50 €
Log in for availability
1 * 10 items
Product Details & Documents
HDPE filter discs for use between filter paper and perforated Büchner plate to prevent clogging of filter holes.
Specifications
  • Ø:
    240 mm
  • Seq:
    08