Tanks
Grey, 40 l, 350×560×304 mm
Catalog # 216-0257
Supplier: KARTELL
Jump to:
Product Details & Documents
Made of HDPE, grey.
- Moulded grips
- Sturdy
Useful general purpose tanks that can be stacked for storage.
Specifications
- Capacity:40 l
- Colour:Grey
- W×D×H:350×560×304 mm