Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Dropping pipettes, 1,5 ml, Ø×L 8×134 mm, 1,5 ml, 8, 134 mm, -
Catalog # 612-1778
Supplier:   KARTELL
Dropping pipettes
Catalog # 612-1778
Supplier:  KARTELL
Dropping pipettes
Dropping pipettes
Dropping pipettes, 1,5 ml, Ø×L 8×134 mm, 1,5 ml, 8, 134 mm, -
Price
45,30 €
Log in for availability
1 * 100 items
Product Details & Documents
PE, graduated.
  • Moulded in one piece
  • High capacity reservoir for take-up and expulsion of liquids
  • Ideal replacement for traditional glass Pasteur pipette and rubber teat
Specifications
  • Ext.Ø:
    8
  • Sterile:
    -
  • Capacity:
    1,5 ml
  • Description:
    Dropping pipettes, 1,5 ml, Ø×L 8×134 mm
  • Length:
    134 mm